Order Entry
Singapore
ContactUsLinkComponent
Bromocresol green 0,04% in aqueous solution
Bromocresol green 0,04% in aqueous solution
Catalog # RICC1200-16
Supplier:  Ricca Chemical
CAS Number:  
undefined
Bromocresol green 0,04% in aqueous solution
Catalog # RICC1200-16
Supplier:  Ricca Chemical
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :698.02 g/mol
Storage Temperature:Ambient
EINECS:200-972-8
CAS Number:76-60-8
Formula:C₂₁H₁₄Br₄O₅S
MDL Number:MFCD00005874