Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Conductivity Standards
Conductivity standard 84 µS/cm (25 °C; KCl)
Conductivity standard 84 µS/cm (25 °C; KCl)
Conductivity standard 84 µS/cm (25 °C; KCl)
Catalog # XXXXECCON84BT
Supplier:
Chemicals
CAS Number:
Expand image
Conductivity standard 84 µS/cm (25 °C; KCl)
Catalog # XXXXECCON84BT
Supplier:
Chemicals
CAS Number:
Specifications
Pack type:
Glass bottle
Pk:
480 mL
Reference temperature:
25 °C
Description:
Conductivity standard 84 µS/cm (KCl)
Specifications
About this item
Product Details
Product Details