Order Entry
Singapore
ContactUsLinkComponent
Sodium hypochlorite 10 - 15% available chlorine in aqueous solution, Reagent Grade
Corrosive
Environment
Sodium hypochlorite 10 - 15% available chlorine in aqueous solution, Reagent Grade
Catalog # HONC425044-1L
CAS Number:  
Corrosive
Environment
Sodium hypochlorite 10 - 15% available chlorine in aqueous solution, Reagent Grade
Catalog # HONC425044-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay of active chlorine:
    10 - 15 %
  • Composition:
    available chlorine, 10-15%

Specification Test Results

About this item

 :
 :74.44 g/mol
Boiling Pt:111 °C
Melting Pt:-30 to -20 °C
Melting Pt:–30…–20 °C
Density:1,206 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-668-3
UN:1791
ADR:8,II
 :GHS05
 :GHS09
CAS Number:7681-52-9
Formula:ClNaO
MDL Number:MFCD00011120