Bromocresol green
Catalog # LOVI511760
Supplier: Lovibond
CAS Number:
Specifications
- Pk:100
- Seq:0100
Specifications
About this item
:
:698.02 g/mol
Merck Index:13,01370
Melting Pt:217…218 °C
Storage Temperature:Ambient
EINECS:200-972-8
CAS Number:76-60-8
Formula:C₂₁H₁₄Br₄O₅S
MDL Number:MFCD00005874