Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Phenyl-5-oxazolone
2-Phenyl-5-oxazolone ≥97%
2-Phenyl-5-oxazolone ≥97%
2-Phenyl-5-oxazolone ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
2-Phenyl-5-oxazolone ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
1199-01-5
:
Formula:
C₉H₇NO₂
MDL Number:
MFCD00014517
:
161.16 g/mol
Melting Pt:
89…92 °C
EINECS:
214-840-2
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAL00194
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation