Order Entry
Singapore
ContactUsLinkComponent
 
Probenecid
Catalog # EDQMP3000000
Supplier:  EDQM
CAS Number:  
Probenecid
Catalog # EDQMP3000000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :285.4 g/mol
EINECS:200-344-3
CAS Number:57-66-9
Formula:C₁₃H₁₉NO₄S
MDL Number:MFCD00038402