Order Entry
Singapore
ContactUsLinkComponent
Conductivity standard 100000 µS/cm (25 °C; 65750 ppm; NaCl)
Conductivity standard 100000 µS/cm (25 °C; 65750 ppm; NaCl)
Catalog # RICC2249-01
Supplier:  Ricca Chemical
CAS Number:  
Conductivity standard 100000 µS/cm (25 °C; 65750 ppm; NaCl)
Catalog # RICC2249-01
Supplier:  Ricca Chemical
CAS Number:  

Specifications

  • Pk:
    1
  • Reference temperature:
    25 °C
  • Description:
    Conductivity standard 100,000 µS/cm (65,750 ppm; NaCl)
  • Conductivity:
    100 000 µS/cm

Specifications

About this item