Conductivity standard 5000 µS/cm (25 °C; 2620 ppm; NaCl) in aqueous solution
Catalog # RICC2245-16
Supplier: Ricca Chemical
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Reference temperature:25 °C
- Description:Conductivity standard 5000 µS/cm (2,620 ppm; NaCl) in aqueous solution
- Conductivity:5000 µS/cm