Conductivity standard 1000 µS/cm (25 °C; 495 ppm; NaCl) in aqueous solution
Catalog # RICC2243-32
Supplier: Ricca Chemical
CAS Number:
Specifications
- Pk:1
- Reference temperature:25 °C
- Description:Conductivity standard 1000 µS/cm (495 ppm; NaCl) in aqueous solution
- Conductivity:1000 µS/cm