
Ethyl-5-amino-3-methyl-1H-pyrazole-4-carboxylate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:23286-70-6
:
Formula:C₇H₁₁N₃O₂
MDL Number:MFCD01210846
:169.18 g/mol
Melting Pt:108 °C
:GHS07
Storage Temperature:Ambient