Order Entry
Singapore
ContactUsLinkComponent
Centrifuge tubes, graduated, conical-bottom, with screw caps, KIMAX®, KIMBLE®
Centrifuge tubes, graduated, conical-bottom, with screw caps, KIMAX®, KIMBLE®
Catalog # KIML45200-10
Supplier:  KIMBLEKONTES
Centrifuge tubes, graduated, conical-bottom, with screw caps, KIMAX®, KIMBLE®
Catalog # KIML45200-10
Supplier:  KIMBLEKONTES

Specifications

  • Closure type:
    Screw cap
  • Packed:
    12/case
  • Thread:
    15-415
  • Capacity:
    10 ml
  • Length:
    116 mm
  • Seq:
    01
  • Pk:
    1

Specifications

About this item

Borosilicate glass, 33 expansion, low extractable.

  • Heavy walled tubes to withstand higher centrifugation speeds
  • Graduated and calibrated to contain
  • Scale is durable white ceramic enamel
  • Top has a screw thread finish
  • Autoclavable cap with rubber liner is included but not attached
Certifications: Glass conforms to USP Type I and ASTM E438, Type I, Class A requirements