Capillary viscometer, Cannon-Fenske, KIMBLE®
Catalog # KIML46460-350
Supplier: KIMBLEKONTES
Specifications
- Range:100 - 500 cSt
- Description:Cannon-Fenske capillary viscometer
- Pk:1
Specifications
About this item
This capillary viscometer requires calibration against a standard liquid of known viscosity or against a second viscometer with a known constant.
- Permanently marked with individual serial number
- Black printed divisions and graduations
- Efflux time greater than 200 seconds
Designed for kinematic viscosities of transparent liquids.
Certifications: ASTM D445 – ASTM D446