Order Entry
Singapore
ContactUsLinkComponent
Shell vials, short style, KIMBLE®
Shell vials, short style, KIMBLE®
Catalog # KIML60931-1
Supplier:  KIMBLEKONTES
undefined
Shell vials, short style, KIMBLE®
Catalog # KIML60931-1
Supplier:  KIMBLEKONTES
Specifications
  • Cap included:
    No
  • Ø×H:
    15×45 mm
  • Capacity:
    4,00 ml
  • Colour:
    Clear
  • Pk:
    1.440
Specifications
About this item
These shell vials are made from clear KIMAX® N-51A borosilicate glass.
  • Plain top design
  • Lab pack quantities