Order Entry
Singapore
ContactUsLinkComponent
Oil dilution distilling receiver, with standard taper joints, PYREX®
Oil dilution distilling receiver, with standard taper joints, PYREX®
Catalog # CORN3582
Supplier:  Corning
undefined
Oil dilution distilling receiver, with standard taper joints, PYREX®
Catalog # CORN3582
Supplier:  Corning
Specifications
  • Cone size:
    24/40 NS
  • Socket size:
    24/40 NS
  • Pk:
    1
Specifications
About this item
Distallation receiver to determine the amount of dilution in crankcase oils when gasoline has been used as fuel. Receiver design keeps heat-sensitive materials away from the hot base of the flask. Receiver has a [ST] 24/40 top outer joint and a [ST] 24/40 sidearm inner joint.
Certifications: Receiver meets ASTM D322 requirements.