Iodine determination flasks, with stopper, PYREX®
: Corning
PYREX® borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Blown in a specially-designed mold to ensure uniformity and proper contour. The stopper handle projects above the liquid seal trough to facilitate removal of the stopper. With No. 22 [ST] stopper. ASTM D29 and D555.