Volumetric flasks, class A, with stopper, PYREX®
Supplier: Corning
Certificates
About this item
PYREX® borosilicate glass.
Flasks feature sharp, permanent graduation line and large white block letter inscriptions. Body is machine-blown with sealed heavy-tubing neck for increased strength. Hollow glass ground NS stoppers, where included, are interchangeable.
All flasks are graduated to Class A volumetric tolerances and calibrated to meet ASTM specifications E-694, E-542, and E-288. Certified flasks are individually serialised and supplied with NIST-traceable certificate of identification and capacity.