Measuring (Mohr) pipettes, color-coded, tempered top and tip, colored markings, PYREX®
Supplier: Corning
Certificates
About this item
These reusable glass pipettes are colour-coded by size for easy identification and sorting.
- Top end is constricted (Mohr type)
- Coloured graduations are enamelled onto the glass
- Calibrated 'to deliver' (TD) their total capacity without blow-out
Sharp, fine lines provide accurate reading. Large numbers are easy to read. The top end of 5 to 25 ml sizes is constricted.
Certifications: ASTM E-1293