Mixing measuring cylinder, tall form, borosilicate glass, PYREX®
: Corning
Borosilicate glass with hollow glass tapered stopper.
- Calibrated to contain (TC, In)
- Hexagonal base
- Ring marks at major graduations, marks and inscriptions in durable white enamel