Burettes, straight PTFE stopcock, PYREX®
Supplier: Corning
Certificates
About this item
Borosilicate glass, class A.
- Red coloured graduations feature permanent markings, fine clear lines
- Large easy to read numbers
- Smooth micro-finish stopcock assures leak-resistant performance
Burette tips are carefully drawn from precision bore tubing to ensure proper drainage rates.
Capacity tolerance is established by ASTM E287 and calibrated in accordance with ASTM E542.
Certifications: Capacity tolerance is established by ASTM E287 and calibrated in accordance with ASTM E542.
Delivery: Burettes are supplied complete with PTFE stopcock and dust cover.