Order Entry
Singapore
ContactUsLinkComponent
Specific gravity bottle, Hubbard-Carmick, PYREX®
Specific gravity bottle, Hubbard-Carmick, PYREX®
Catalog # CORN1620-25
Supplier:  Corning
undefined
Specific gravity bottle, Hubbard-Carmick, PYREX®
Catalog # CORN1620-25
Supplier:  Corning
Specifications
  • Volume (nom.):
    25 ml
  • Pk:
    1
Specifications
About this item
For viscous fluids, semi-solid bitumens, and emulsions according to ASTM D70. Includes NS 24/12 stopper.