Order Entry
Singapore
ContactUsLinkComponent
 
Hypering bottle, graduated, PYREX®
  CORN1350-2L
 :  Corning
undefined
Hypering bottle, graduated, PYREX®
  CORN1350-2L
 :  Corning

 

  • Ext.Ø:
    120 mm
  • Capacity:
    2000 ml
  • Height:
    296 mm
  • Neck Ø ext.:
    54 mm
  • Pk:
    1

 

 

Cylindrical, wide neck, borosilicate bottle has a tooled neck and graduations that can be read from top or bottom. They fit a no. 10 rubber stopper (not included).

  • Very good chemical resistance
  • Very good temperature resistance