Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cupri-citric solution
Cupri-citric solution R1 Reag. Ph. Eur. 1023200
Cupri-citric solution R1 Reag. Ph. Eur. 1023200
Catalog # VWRC87813.290
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Expand image
Cupri-citric solution R1 Reag. Ph. Eur. 1023200
Catalog # VWRC87813.290
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Specifications
Pack type:
Plastic bottle
Pk:
1 L
Seq:
5037
Specifications
About this item
Product Details
Documentation
:
Product Details
Documentation