$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
4-Bromo-4'-hydroxybiphenyl
4-Bromo-4'-hydroxybiphenyl ≥98%
4-Bromo-4'-hydroxybiphenyl ≥98%
4-Bromo-4'-hydroxybiphenyl ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
4-Bromo-4'-hydroxybiphenyl ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
29558-77-8
:
Formula:
C₁₂H₉BrO
MDL Number:
MFCD00059076
:
249.11 g/mol
Melting Pt:
163…168 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAA10819
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation