$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
Enzymes
Pig Desoxyribonuclease II (from Spleen)
Pig Desoxyribonuclease II (from Spleen)
ALFAJ63389.DX
:
Thermo Fisher Scientific
:
Pig Desoxyribonuclease II (from Spleen)
ALFAJ63389.DX
:
Thermo Fisher Scientific
:
Pk:
80 KU
CAS No.:
9025-64-3
Species:
Pig
Enzyme name:
Desoxyribonuclease II
Enzyme synonyms:
DNase II