Bismuth vanadium tetraoxide ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14059-33-7
:
Formula:BiO₄V
MDL Number:MFCD00064836
:323.00 g/mol
Storage Temperature:Ambient
EINECS:237-898-0
UN:3285
ADR:6.1,III
:GHS07