Order Entry
Singapore
ContactUsLinkComponent
 
Health
Corrosive
Bromotris(dimethylamino)phosphonium hexafluorophosphate ≥98%
Catalog # ALFAH26708.06
CAS Number:  
Health
Corrosive
Bromotris(dimethylamino)phosphonium hexafluorophosphate ≥98%
Catalog # ALFAH26708.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

CAS Number:50296-37-2
 :
Formula:C₆H₁₈BrF₆N₃P₂
MDL Number:MFCD00191864
 :388.07 g/mol
Melting Pt:>300 °C
ADR:8,II
 :GHS08
 :GHS05