Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Bromo-2-hydroxybenzophenone
5-Bromo-2-hydroxybenzophenone ≥97%
5-Bromo-2-hydroxybenzophenone ≥97%
5-Bromo-2-hydroxybenzophenone ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
5-Bromo-2-hydroxybenzophenone ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
55082-33-2
:
Formula:
C₁₃H₉BrO₂
MDL Number:
MFCD00525062
:
277.11 g/mol
Melting Pt:
108…111 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAB25507
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation