4-Bromo-3,5-dimethyl-1H-pyrazole ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3398-16-1
:
Formula:C₅H₇BrN₂
MDL Number:MFCD00005242
:175.03 g/mol
Melting Pt:118…123 °C
Storage Temperature:Ambient
EINECS:222-263-2