
3-Amino-4-bromo-5-methyl-1H-pyrazole hydrobromide 98%
Catalog # ALFAA14487.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:256.94 g/mol
Melting Pt:178…181 °C
:GHS07
Storage Temperature:Ambient
CAS Number:167683-86-5
Formula:C₄H₇Br₂N₃
MDL Number:MFCD00053023