
Forskolin (from Coleus forskohlii)
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:66575-29-9
:
Formula:C₂₂H₃₄O₇
MDL Number:MFCD00082317
:410.51 g/mol
Melting Pt:228…230 °C (lit.)
EINECS:266-410-9
:GHS07
Storage Temperature:Freezer