D(-)-Luciferin sodium salt ≥98.5%
Catalog # APLIA1006.0010
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:302.30 g/mol
CAS Number:103404-75-7
Formula:C₁₁H₇N₂NaO₃S₂
MDL Number:MFCD00044938