Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Buffers
Biological Buffers
Buffer, sodium phosphate, 1M solution, pH 7
Buffer, sodium phosphate, 1M solution, pH 7
Buffer, sodium phosphate, 1M solution, pH 7
Catalog # TEKOP2070
Supplier:
TEKNOVA
CAS Number:
Expand image
Buffer, sodium phosphate, 1M solution, pH 7
Catalog # TEKOP2070
Supplier:
TEKNOVA
CAS Number:
Specifications
Pack type:
Glass bottle
Pk:
1 L
Description:
Sodium phosphate buffer solution pH 7 (1 mol/l)
Specifications
About this item
Product Details
Product Details