

Diclofenac sodium salt
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15307-79-6
:
Formula:C₁₄H₁₀Cl₂NNaO₂
MDL Number:MFCD00082251
:318.1 g/mol
Merck Index:14,3081
Melting Pt:283…285 °C
Storage Temperature:Ambient
EINECS:239-346-4
UN:2811
ADR:6.1,III
:GHS06
:GHS08