Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-(6-Methoxy-3-pyridyl)thiourea
N-(6-Methoxy-3-pyridyl)thiourea ≥97%
N-(6-Methoxy-3-pyridyl)thiourea ≥97%
:
Thermo Fisher Scientific
N-(6-Methoxy-3-pyridyl)thiourea ≥97%
:
Thermo Fisher Scientific
:
:
C₇H₉N₃OS
:
MFCD10695141
:
183.23 g/mol
:
190…194 °C
:
GHS06
:
GHS07
:
Ambient
msds_ALFAH33009
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years