Order Entry
Singapore
ContactUsLinkComponent
N-(6-Bromo-2-pyridyl)thiourea ≥97%
N-(6-Bromo-2-pyridyl)thiourea ≥97%
Catalog # ALFAH34220.MD
CAS Number:  
N-(6-Bromo-2-pyridyl)thiourea ≥97%
Catalog # ALFAH34220.MD
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

CAS Number:439578-83-3
 :
Formula:C₆H₆BrN₃S
MDL Number:MFCD22380711
 :232.10 g/mol
Melting Pt:217…221 °C
 :GHS06
 :GHS07
Storage Temperature:Ambient