Order Entry
Singapore
ContactUsLinkComponent
N-(3-Bromo-2-pyridyl)thiourea ≥97%
N-(3-Bromo-2-pyridyl)thiourea ≥97%
Catalog # ALFAH33465.03
CAS Number:  
N-(3-Bromo-2-pyridyl)thiourea ≥97%
Catalog # ALFAH33465.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Formula:C₆H₆BrN₃S
MDL Number:MFCD22380708
 :232.10 g/mol
Melting Pt:160…166 °C
 :GHS06
 :GHS07
Storage Temperature:Ambient