Order Entry
Singapore
ContactUsLinkComponent
Bottle top vacuum filtration systems
Bottle top vacuum filtration systems
Catalog # BIBB5810/3
Supplier:  SciLabware
undefined
Bottle top vacuum filtration systems
Catalog # BIBB5810/3
Supplier:  SciLabware
Specifications
  • Description:
    Support base with sintered glass disc and vacuum connection, for 47 mm membrane
  • Seq:
    03
  • Pk:
    1
Specifications
About this item
This filteration system is used for microbiology and preparation of HPLC solvents. It is used with 47 mm Ø membranes.
  • Borosilicate glass sintered disc to support membrane
  • Each component is also available to order separately