Order Entry
Singapore
ContactUsLinkComponent
Gel filtration chromatography columns, Tricorn™ Superose™ 12 GL
Gel filtration chromatography columns, Tricorn™ Superose™ 12 GL
Catalog # GEHE17-5173-01
Supplier:  Cytiva
undefined
Gel filtration chromatography columns, Tricorn™ Superose™ 12 GL
Catalog # GEHE17-5173-01
Supplier:  Cytiva

Specifications

  • Flow rate:
    0,5 - 1 ml/min
  • Particle Size:
    11 µm
  • Max. pressure:
    30 bar
  • Capacity:
    24 ml
  • Description:
    Superose™ 12 10/300 GL
  • Seq:
    01
  • pH range:
    3 - 12
  • Column hardware:
    Glass
  • Bed dimensions:
    10×300 mm
  • Pk:
    1

Specifications

About this item

Superose™ 12 10/300 GL are pre-packed Tricorn™ columns for high performance gel filtration with a broad protein fractionation range of 1000 to 300000.

  • Easy, efficient scale up from pre-packed Superose™ 10/300 GL columns to Superose™ prep grade lab packs
  • Low ionic strength eluents (<0,05 M) can utilise a hydrophobic interaction component to alter the selectivity of Superose™ for some lipids, peptides and small aromatic compounds
  • Pre-packed Tricorn™ columns are durable, high performance glass columns for reliable and reproducible separations