Order Entry
Singapore
Orders LinkContactUsLinkComponent
Magnetic stirring bars, PTFE, octagonal
Magnetic stirring bars, PTFE, octagonal
Catalog #
Magnetic stirring bars, PTFE, octagonal
Catalog #
Supplier:  
Magnetic stirring bars, PTFE, octagonal
Magnetic stirring bars, PTFE, octagonal
Product Details & Documents
magnetic stiirring bars with center rings and permanent magnet AlNiCo V core. The eight-sided shape gives rise to significant turbulence, and thus achieves effective mixing, even at low spin rates. The middle ring also promotes stable centering with convex or uneven bottoms.
  • Octagonal shape gives rise to significant turbulence, and thus achieves effective mixing, even at low spin rates
  • Cerntre ring also promotes stable centering with convex or uneven bases