Methotrexate
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Synonyms: (S)-2-(4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzamido)pentanedioic acid
- CAS Number:59-05-2
- Formula:C₂₀H₂₂N₈O₅
- MDL Number:MFCD00150847
- Molecular Weight:454.44 g/mol
- Merck Index:14,5985
- Storage Temperature:Freezer
- EINECS:200-413-8
- UN:1544
- ADR:6.1,III
2 Options Available Below
Product Details & Documents
Synonyms: (S)-2-(4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzamido)pentanedioic acid
- CAS Number:59-05-2
- Formula:C₂₀H₂₂N₈O₅
- MDL Number:MFCD00150847
- Molecular Weight:454.44 g/mol
- Merck Index:14,5985
- Storage Temperature:Freezer
- EINECS:200-413-8
- UN:1544
- ADR:6.1,III
Documents
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



