Bestatin hydrochloride ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride) ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:65391-42-6
:
Formula:C₁₆H₂₅ClN₂O₄
MDL Number:MFCD00058004
:308.4 · 36.5 g/mol
Melting Pt:216…218 °C
Storage Temperature:Ambient