Vitamin A palmitate, 1.7 MIU/g
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:79-81-2
:
Formula:C₃₆H₆₀O₂
MDL Number:MFCD00019414
:524.87 g/mol
Merck Index:14,10013
Melting Pt:28…29 °C
EINECS:201-228-5
:GHS08
:GHS07
Storage Temperature:Refrigerator