


Toluene
Catalog # BJAA65752
Supplier: BURDICK JACKSON
CAS Number:
Specifications
- Pk:185 L
- Seq:5491
Specifications
About this item
:
:92.14 g/mol
Merck Index:13,09607
Boiling Pt:110,6 °C (1013 hPa)
Melting Pt:–95 °C
Density:0,867 g/cm³ (20 °C)
Flash Pt:4 °C
Storage Temperature:Ambient
EINECS:203-625-9
UN:1294
ADR:3,II
REACH:05-2114615130-69
:GHS02
:GHS08
:GHS07
CAS Number:108-88-3
Formula:C₆H₅CH₃
MDL Number:MFCD00008512