Barium sulphate 97.5-100.5% USP, Multi-Compendial, J.T.Baker®
Catalog # BAKR1041-01
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Poly bottle, white, blue T/E cap
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- GMP Manufactured Product:Passes Test
- Meets B.P. Chemical Specifications:Passes Test
- Meets E.P. Chemical Specifications:Passes Test
- Meets U.S.P Requirements:3.5 - 10.0
- CAUTION: For Manufacturing, processing or repackaging:≤0.5
- Bulk Pharmaceutical Chemical:≤0.3%
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Bulkiness:Passes Test
- USP - pH of 10% Aqueous Solution at 25°C:3.5 - 10.0
- USP - Sulfide (µg/g):≤ 0.5
- USP - Acid-Soluble Substances:≤ 0.3 %
- USP - Soluble Barium Salts:≤ 0.001 %
- USP - Assay:97.5 - 100.5 %
- USP - Particulate Matter:Passes Test
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Acidity or Alkalinity:Passes Test
- EP/BP - Acid-Soluble Substances:≤ 0.3 %
- EP/BP - Oxidizable Sulfur Compounds:Passes Test
- EP/BP - Soluble Barium Salts:≤ 10 ppm
- EP/BP - Phosphate (PO₄):≤ 50 ppm
- EP/BP - Arsenic (As):≤ 2 ppm
- EP/BP - Loss on Ignition:≤ 2.0 %
- EP/BP - Sediment:Passes Test
Specification Test Results
About this item
:
:233.39 g/mol
Merck Index:13,00997
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:1345 °C
Density:4,5 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-784-4
CAS Number:7727-43-7
Formula:BaSO₄
MDL Number:MFCD00003455