

Thioacetamide ≥99.0%, crystals, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
BAKR8984-01
: Avantor
:
- Pack type:Glass bottle, amber, wide mouth, black T/E cap
- Pk:500 g
- Seq:3645
- For Laboratory,Research,or Manufacturing Use:= 99.0 %
- Assay (CH₃CSNH₂):≥99.0 %
- Melting Point:111.0 - 114.0 °C
- Residue after Ignition:≤0.050 %
:
:75.13 g/mol
:13,09391
:113…114 °C
:1,336 g/cm³ (20 °C)
:200-541-4
:GHS08
:GHS07
:62-55-5
:CH₃CSNH₂
:MFCD00008070