$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
3-Nitrobenzylamine hydrochloride
3-Nitrobenzylamine hydrochloride 97%
3-Nitrobenzylamine hydrochloride 97%
:
Thermo Fisher Scientific
3-Nitrobenzylamine hydrochloride 97%
:
Thermo Fisher Scientific
:
26177-43-5
:
:
C₇H₉ClN₂O₂
:
MFCD00012858
:
188.62 g/mol
:
∼ 225 °C (dec.)
:
247-502-8
:
GHS07
:
Ambient
msds_ALFAB23349
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years