Ethyl-3-hydroxyhexanoate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2305-25-1
:
Formula:C₈H₁₆O₃
MDL Number:MFCD00036604
:160.21 g/mol
Boiling Pt:90…92 °C (14 mmHg)
Density:0,974
Flash Pt:94 °C (202 °F)
Storage Temperature:Ambient
EINECS:218-973-7