SPE cartridges, (Na₂SO₄) CHROMAFIX® Dry
Supplier: MACHEREY-NAGEL
Certificates
About this item
CHROMAFIX® Dry is a special phase for drying organic samples.
- Polypropylene cartridges in three different sizes with different adsorbent weights (S: 0,4 ml, M: 0,8 ml, L: 1,8 ml)
- Anhydrous high-purity sodium sulfate which forms Glauber’s salt with traces of water
Application: Connected with a syringe via Luer fittings traces of water from organic solutions will be removed. For removal of larger quantities of water several cartridges can be combined in series.