2-Acetyl-5-nitrothiophene
Catalog # PFALN12568.1G
Supplier: PFALTZ BAUER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
UN:2811
ADR:6.1,III
CAS Number:39565-00-9
Formula:C₆H₅NO₃S