Order Entry
Singapore
ContactUsLinkComponent
Magnesium sulphate 0.1 M volumetric solution, Fluka™
Magnesium sulphate 0.1 M volumetric solution, Fluka™
Catalog # HONC34276-1L
CAS Number:  
undefined
Magnesium sulphate 0.1 M volumetric solution, Fluka™
Catalog # HONC34276-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002

Specification Test Results

About this item

 :
 :120.37 g/mol
Storage Temperature:Ambient
EINECS:231-298-2
CAS Number:7487-88-9
Formula:MgSO₄
MDL Number:MFCD00011110